missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene, 99+%

Catalog No. AAH2777609
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2777609 10 g
AAH2777603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2777609 Supplier Thermo Scientific Chemicals Supplier No. H2777609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 80221-11-0 | C22H26 | 290.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80221-11-0
Molecular Formula C22H26
Molecular Weight (g/mol) 290.45
MDL Number MFCD04038794
InChI Key UUVMZCQRPVPWNI-UHFFFAOYSA-N
Synonym 1-n-butyl-4-4-butylphenyl ethynyl benzene, 1,2-bis 4-butylphenyl ethyne, 1-butyl-4-2-4-butylphenyl ethynyl benzene, bis 4-butylphenyl ethyne, 1,2-bis 4-n-butylphenyl acetylene, 1,2-di-4-n-butylphenyl acetylene, 1,1'-ethyne-1,2-diyl bis 4-butylbenzene, 1-n-butyl-4-4-butylphenyl ethynyl benzene, 99+%
PubChem CID 13710203
IUPAC Name 1-butyl-4-[2-(4-butylphenyl)ethynyl]benzene
SMILES CCCCC1=CC=C(C=C1)C#CC1=CC=C(CCCC)C=C1

Specifications

Melting Point 40°C to 42°C
Boiling Point 172°C to 174°C (0.5 mmHg)
Quantity 10 g
Formula Weight 290.45
Percent Purity ≥99%
Chemical Name or Material 1-n-Butyl-4-[(4-butylphenyl)ethynyl]benzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only