missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrazole-5-methanol 98.0+%, TCI America™

Catalog No. M2831200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2831200MG 200 mg
M28311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2831200MG Supplier TCI America Supplier No. M2831200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84547-61-5
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD08556301
InChI Key WQFOGLYQVFBDEY-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazol-5-yl methanol, 5-hydroxymethyl-1-methyl-1h-pyrazole, 1h-pyrazole-5-methanol, 1-methyl, 2-methylpyrazol-3-yl methanol, 5-hydroxymethyl-1-methylpyrazole, 1h-pyrazole-5-methanol,1-methyl, 1-methylpyrazol-5-yl methan-1-ol, 1-methylpyrazole-5-methanol, 1-methyl-1h-pyrazol-5-yl ;methanol
PubChem CID 11815316
IUPAC Name (2-methylpyrazol-3-yl)methanol
SMILES CN1C(=CC=N1)CO

Specifications

Melting Point 58°C
Color White
Quantity 200 mg
Formula Weight 112.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrazole-5-methanol