missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopropane-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. M08855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08855G 5 g
M088525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08855G Supplier TCI America Supplier No. M08855G
Only null left

Chemical Identifiers

CAS 6914-76-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00001290
InChI Key DIZKLZKLNKQFGB-UHFFFAOYSA-N
Synonym 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa
PubChem CID 81326
IUPAC Name 1-methylcyclopropane-1-carboxylic acid
SMILES CC1(CC1)C(=O)O

Specifications

Color White-Yellow
Boiling Point 185°C
Quantity 5 g
UN Number 3261
Formula Weight 100.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Methylcyclopropane-1-carboxylic Acid