missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl L-Glutamate 98.0+%, TCI America™

Catalog No. M18615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18615G Supplier TCI America Supplier No. M18615G

Specifications

CAS 6384-08-3
Melting Point 145°C
Color White
Molecular Formula C6H11NO4
Quantity 5 g
MDL Number MFCD00171677
Synonym h-glu-ome, s-4-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid 1-methyl ester, l-glutamate methylester, 1-methyl l-glutamate, l-glutamate methyl ester, glutamic acid methyl ester, 4s-4-amino-5-methoxy-5-oxopentanoic acid, gluome, glu alpha-ome-oh
InChI Key SEWIYICDCVPBEW-UHFFFAOYNA-N
SMILES COC(=O)C(CCC(=O)O)N
IUPAC Name (4S)-4-amino-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol) 161.16
PubChem CID 440550
ChEBI CHEBI:28575
Formula Weight 161.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl L-Glutamate
Show More Show Less
EINECSNumber (9)-0418
TSCA No