missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 921939-08-4
Molecular Formula C12H9F3N2O3
Molecular Weight (g/mol) 286.21
MDL Number MFCD06203565
InChI Key JLFWKPKAZSVCKI-UHFFFAOYSA-N
Synonym 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carboxylic acid
PubChem CID 24229721
IUPAC Name 1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carboxylic acid
SMILES CN1N=C(C(C(O)=O)=C1OC1=CC=CC=C1)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC64801CB
SDS View Documents
Thermo Scientific Chemicals
CC64801CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64801DA
SDS View Documents
Thermo Scientific Chemicals
CC64801DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

921939-08-4
286.21
JLFWKPKAZSVCKI-UHFFFAOYSA-N
24229721
CN1N=C(C(C(O)=O)=C1OC1=CC=CC=C1)C(F)(F)F
C12H9F3N2O3
MFCD06203565
1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carboxylic acid
1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carboxylic acid

Specifications

921939-08-4
100.0
Amber Glass Bottle
MFCD06203565
1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carboxylic acid
CN1N=C(C(C(O)=O)=C1OC1=CC=CC=C1)C(F)(F)F
286.21
286.21
1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
96.5
97%
C12H9F3N2O3
250mg
JLFWKPKAZSVCKI-UHFFFAOYSA-N
1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carboxylic acid
24229721
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'