missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 109925-42-0
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD01936001
InChI Key JDLPKSIFGPLXAU-UHFFFAOYSA-N
Synonym 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenoxy-3-trifluoromethyl, acmc-20a3tk, methylphenoxytrifluoromethylpyrazolecarbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxyaldehyde
PubChem CID 2775493
IUPAC Name 1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carbaldehyde
SMILES CN1C(=C(C(=N1)C(F)(F)F)C=O)OC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC64804CB
SDS View Documents
Thermo Scientific Chemicals
CC64804CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC64804DA
SDS View Documents
Thermo Scientific Chemicals
CC64804DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

109925-42-0
270.211
JDLPKSIFGPLXAU-UHFFFAOYSA-N
2775493
CN1C(=C(C(=N1)C(F)(F)F)C=O)OC2=CC=CC=C2
C12H9F3N2O2
MFCD01936001
1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenoxy-3-trifluoromethyl, acmc-20a3tk, methylphenoxytrifluoromethylpyrazolecarbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxyaldehyde
1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carbaldehyde

Specifications

109925-42-0
100.0
Amber Glass Bottle
MFCD01936001
1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl pyrazole-4-carbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenoxy-3-trifluoromethyl, acmc-20a3tk, methylphenoxytrifluoromethylpyrazolecarbaldehyde, 1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazole-4-carboxyaldehyde
CN1C(=C(C(=N1)C(F)(F)F)C=O)OC2=CC=CC=C2
270.211
270.21
1-Methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
96.5
97%
C12H9F3N2O2
250mg
JDLPKSIFGPLXAU-UHFFFAOYSA-N
1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazole-4-carbaldehyde
2775493
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents