missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline, 97%, Thermo Scientific™

Catalog No. p-3586609
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58739CB 250mg Amber Glass Bottle
CC58739DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC58739CB Supplier Thermo Scientific Chemicals Supplier No. CC58739CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 934570-43-1
Molecular Formula C15H22BNO2
Molecular Weight (g/mol) 259.156
MDL Number MFCD09879913
InChI Key CZYIGZLFKRAXMV-UHFFFAOYSA-N
Synonym 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline, 1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole, 1-methylindoline-5-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1-methylindolin-5-yl-1,3,2-dioxaborolane, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole
PubChem CID 24229591
IUPAC Name 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(CC3)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 259.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.