missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 405.33 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Methyl-3-propylimidazolium Bis(trifluoromethanesulfonyl)imide |
Safety and Handling
| TSCA | No |