missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Numéro de catalogue. p-3175603
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC46601CB 250mg Amber Glass Bottle
CC46601DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC46601CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC46601CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 10250-64-3
Formule moléculaire C11H10N2O2
Poids moléculaire (g/mol) 202.213
Numéro MDL MFCD08271939
Clé InChI RTBQZMPYPFQBHA-UHFFFAOYSA-N
Synonyme 1-methyl-3-phenyl-1h-pyrazole-5-carboxylic acid, 2-methyl-5-phenyl-2h-pyrazole-3-carboxylic acid, 1-methyl-3-phenylpyrazole-5-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1-methyl-3-phenyl, 5-carboxy-1-methyl-3-phenyl-1h-pyrazole, 1-methyl-3-phenyl-1h-5-pyrazolecarboxylic acid, 1h-pyrazole-5-carboxylicacid, 1-methyl-3-phenyl, 1-methyl-3-phenyl-1 h-pyrazole-5-carboxylic acid
PubChem CID 7537625
Nom de l’IUPAC 2-methyl-5-phenylpyrazole-3-carboxylic acid
SOURIRES CN1C(=CC(=N1)C2=CC=CC=C2)C(=O)O

Spécifications

Nom ou substance chimique 1-Methyl-3-phenyl-1H-pyrazole-5-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 202.21
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.