missing translation for 'onlineSavingsMsg'
Learn More

1-méthyl-3-phényl-1H-pyrazole-5-carbaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. CC46604DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC46604DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC46604DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC46604DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 864068-96-2
Formule moléculaire C11H10N2O
Poids moléculaire (g/mol) 186.21
Numéro MDL MFCD08271941
Clé InChI XFGHBJQGDDYIKS-UHFFFAOYSA-N
Synonyme 1-methyl-3-phenyl-1h-pyrazole-5-carbaldehyde, 1-methyl-3-phenylpyrazole-5-carbaldehyde, 2-methyl-5-phenyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-3-phenyl-1h-pyrazole, 1h-pyrazole-5-carboxaldehyde,1-methyl-3-phenyl
PubChem CID 7537629
SOURIRES CN1N=C(C=C1C=O)C1=CC=CC=C1

Spécifications

Nom ou substance chimique 1-Methyl-3-phenyl-1H-pyrazole-5-carbaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Nom de l’IUPAC 2-méthyl-5-phénylpyrazole-3-carbaldehyde
Poids de formule 186.21
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.