missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. M24405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24405G 5 g
M244025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24405G Supplier TCI America Supplier No. M24405G
Only null left

Chemical Identifiers

CAS 403842-84-2
Molecular Formula C13H23F3N2O3S
Molecular Weight (g/mol) 344.39
MDL Number MFCD06411717
InChI Key BCVMAYRUJBXMAP-UHFFFAOYSA-M
Synonym 1-methyl-3-octylimidazolium trifluoromethanesulfonate, 1-methyl-3-n-octylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium triflate, dsstox_cid_27922, dsstox_rid_82675, dsstox_gsid_47946, 1-methyl-3-octylimidazolium triflate, 1-methyl-3-n-octylimidazolium triflate, 1-methyl-3-octylimidazolium trifluoromet
PubChem CID 16212746
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate
SMILES [O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 344.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Trifluoromethanesulfonate