missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%

Catalog No. AAH2716106
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2716106 5 g
AAH2716118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2716106 Supplier Thermo Scientific Chemicals Supplier No. H2716106

CAS: 244193-52-0 | C12H23BF4N2 | 282.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 244193-52-0
Molecular Formula C12H23BF4N2
Molecular Weight (g/mol) 282.13
MDL Number MFCD03095434
InChI Key GXZCAMSPWNHTAE-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium tetrafluoroborate, 1-methyl-3-n-octylimidazolium tetrafluoroborate, 3-methyl-1-octylimidazolium tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, omimbf4, dsstox_cid_27919, dsstox_rid_82672, dsstox_gsid_47943, 3-methyl-1-octylimidazoliumtetrafluoroborate, 1-methyl-3-octylimidazolium tetrafluoroborate hplc
PubChem CID 11460221
SMILES F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Density 1.12
Quantity 5 g
IUPAC Name 1-methyl-3-octylimidazol-1-ium;tetrafluoroborate
Formula Weight 282.13
Percent Purity 99%
Chemical Name or Material 1-Methyl-3-n-octylimidazolium tetrafluoroborate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only