missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-[6-(methylsulfinyl)hexyl]imidazolium p-Toluenesulfonate 97.0+%, TCI America™

Catalog No. p-7133129
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22741G 1 g
M22745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22741G Supplier TCI America Supplier No. M22741G

Chemical Identifiers

CAS 1352947-66-0
Molecular Formula C18H28N2O4S2
Molecular Weight (g/mol) 400.55
MDL Number MFCD27976828
InChI Key YITFXJWLFMQQRC-UHFFFAOYNA-M
PubChem CID 56833668
IUPAC Name 1-(6-methanesulfinylhexyl)-3-methyl-1H-imidazol-3-ium 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1=CN(CCCCCCS(C)=O)C=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 400.55
Percent Purity ≥97.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-[6-(methylsulfinyl)hexyl]imidazolium p-Toluenesulfonate