missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-5-carbaldehyde, 97%, Thermo Scientific™

Numéro de catalogue. CC62304DA
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC62304DA 1g Amber Glass Bottle
CC62304CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC62304DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC62304DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 27258-33-9
Formule moléculaire C5H6N2O
Poids moléculaire (g/mol) 110.12
Numéro MDL MFCD03419801
Clé InChI RAJRANFZSWDUJZ-UHFFFAOYSA-N
Synonyme 1-methyl-1h-pyrazole-5-carbaldehyde, 2-methyl-2h-pyrazole-3-carbaldehyde, 5-formyl-1-methyl-1h-pyrazole, 1-methylpyrazole-5-carboxaldehyde, 5-formyl-1-methylpyrazole, 1-methylpyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1-methyl, 1-methyl-1h-pyrazole-5-carboxaldehyde, pubchem16289, acmc-209gw7
PubChem CID 7019414
Nom de l’IUPAC 2-methylpyrazole-3-carbaldehyde
SOURIRES CN1N=CC=C1C=O

Spécifications

Nom ou substance chimique 1-Methyl-1H-pyrazole-5-carbaldehyde
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 110.12
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.