missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-pyrazole-3-carbonyl chloride, 97%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 1-Methyl-1H-pyrazole-3-carbonyl chloride |
| CAS % minimum | 96.5 |
| CAS % maximal | 100.0 |
| Fourchette de pourcentages d’essai | 95% |
| Emballage | Amber Glass Bottle |
| Quantité | 250mg |
| Poids de formule | 144.56 |
| Pourcentage de pureté | 97% |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. Contact with water liberates toxic gas. Reacts violently with water. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Store in a dry place. Store in a closed container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |