missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-1H-pyrazol-5-yl)methanol, 97%, Thermo Scientific™

Catalog No. CC62309CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC62309CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC62309CB Supplier Thermo Scientific Chemicals Supplier No. CC62309CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 84547-61-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H8N2O
Quantity 250mg
MDL Number MFCD08556301
Synonym 1-methyl-1h-pyrazol-5-yl methanol, 5-hydroxymethyl-1-methyl-1h-pyrazole, 1h-pyrazole-5-methanol, 1-methyl, 2-methylpyrazol-3-yl methanol, 5-hydroxymethyl-1-methylpyrazole, 1h-pyrazole-5-methanol,1-methyl, 1-methylpyrazol-5-yl methan-1-ol, 1-methylpyrazole-5-methanol, 1-methyl-1h-pyrazol-5-yl ;methanol
InChI Key WQFOGLYQVFBDEY-UHFFFAOYSA-N
SMILES CN1C(=CC=N1)CO
IUPAC Name (2-methylpyrazol-3-yl)methanol
Molecular Weight (g/mol) 112.132
PubChem CID 11815316
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.