missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-3-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 4002-83-9
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD08060478
InChI Key KYDLGYCLCFKUHY-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
PubChem CID 7164545
IUPAC Name 1-methylindazole-3-carbaldehyde
SMILES CN1N=C(C=O)C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC06304CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC06304DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

4002-83-9
160.18
KYDLGYCLCFKUHY-UHFFFAOYSA-N
7164545
CN1N=C(C=O)C2=CC=CC=C12
C9H8N2O
MFCD08060478
1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
1-methylindazole-3-carbaldehyde

Specifications

4002-83-9
100.0
C9H8N2O
1-methyl-1h-indazole-3-carbaldehyde, 1-methyl-1h-indazole-3-carboxaldehyde, 3-formyl-1-methyl-1h-indazole, 1-methyl-3-formylindazole, 1h-indazole-3-carboxaldehyde,1-methyl
CN1N=C(C=O)C2=CC=CC=C12
160.18
160.18
1-Methyl-1H-indazole-3-carbaldehyde
96.5
97%
MFCD08060478
KYDLGYCLCFKUHY-UHFFFAOYSA-N
1-methylindazole-3-carbaldehyde
7164545
97%
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents