missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-1,2,3-benzotriazole-5-carbaldehyde, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Quantity | 250mg |
| Formula Weight | 161.16 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Methyl-1H-1, 2, 3-benzotriazole-5-carbaldehyde |