missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1-cyclohexene, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Density | 0.81 |
| Boiling Point | 108°C to 112°C |
| Flash Point | −1°C (30°F) |
| Refractive Index | 1.45 |
| Quantity | 5 g |
| UN Number | UN3295 |
| Beilstein | 1304483 |
| Formula Weight | 96.17 |
| Percent Purity | 96% |
| Chemical Name or Material | 1-Methyl-1-cyclohexene |
Sécurité et manipulation
| Hazard Category | H225-H304-H315-H320-H335 |
| Hazard Statement | GHS H Statement H225-H304 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P302+P352-P305+P351+P338-P312-P331-P332+P313-P362-P372-P374-P380-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S. |
| EINECSNumber | 209-718-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only