missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1,2-epoxycyclohexane 95.0+%, TCI America™

Catalog No. M19835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19835G 5 g
M19831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19835G Supplier TCI America Supplier No. M19835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1713-33-3
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD11846121
InChI Key FIEKVYPYFQSFTP-UHFFFAOYNA-N
Synonym 1,2-Epoxy-1-methylcyclohexane, 1-Methyl-7-oxabicyclo[4.1.0]heptane
PubChem CID 224243
IUPAC Name 1-methyl-7-oxabicyclo[4.1.0]heptane
SMILES CC12CCCCC1O2

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 1993
Formula Weight 112.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methyl-1,2-epoxycyclohexane