missing translation for 'onlineSavingsMsg'
Learn More

1-Methoxy-2-propanol, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019156
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019156 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019156 Supplier Merck Emd Millipore Supplier No. 656925MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Environmental

Specifications

CAS 107-98-2
Ignition Point 532°F
Vapor Pressure 10.9 mmHg (25°C)
Vapor Density 3.12 (vs. air)
Density 0.916 g/mL (at 25°C)
Flash Point 82°C - closed cup
Physical Form Neat
Linear Formula CH3 CH(OH)CH2 OCH3
MDL Number MFCD00004537
Boiling Point 118°C to 119°C
Synonym Propylene glycol methyl ether; Propyleneglycol monomethyl ether
Type Analytical Standard
Quantity 5 mL
Formula Weight 90.12
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less