missing translation for 'onlineSavingsMsg'
Learn More

1-Isocyanato-2-(phenoxymethyl)benzene, 97%, Thermo Scientific™

Numéro de catalogue. CC58506CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC58506CB 250mg Amber Glass Bottle
CC58506DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC58506CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC58506CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 910037-00-2
Formule moléculaire C14H11NO2
Poids moléculaire (g/mol) 225.25
Numéro MDL MFCD09025892
Clé InChI FGEXXDXHIUWRRS-UHFFFAOYSA-N
Synonyme 1-isocyanato-2-phenoxymethyl benzene, 2-phenoxymethyl benzenisocyanate
PubChem CID 18525894
Nom de l’IUPAC 1-isocyanato-2-(phenoxymethyl)benzene
SOURIRES O=C=NC1=CC=CC=C1COC1=CC=CC=C1

Spécifications

Nom ou substance chimique 1-Isocyanato-2-(phenoxymethyl)benzene
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 225.25
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.