missing translation for 'onlineSavingsMsg'
Learn More

1-Iodooctane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. I0321500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0321500G 500 g
I032125G 25 g
I0321100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0321500G Supplier TCI America Supplier No. I0321500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-27-6
Molecular Formula C8H17I
Molecular Weight (g/mol) 240.128
MDL Number MFCD00001105
InChI Key UWLHSHAHTBJTBA-UHFFFAOYSA-N
Synonym n-octyl iodide, octyl iodide, octane, 1-iodo, 1-octyl iodide, 1-n-octyl iodide, 1-jodoktan, 1-jodoktan czech, 1-iodooctane;n-octyl iodide, iodoctane, iodooctane
PubChem CID 12380
IUPAC Name 1-iodooctane
SMILES CCCCCCCCI

Specifications

Color Red-Yellow
Boiling Point 226°C
Quantity 500 g
Formula Weight 240.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodooctane (stabilized with Copper chip)
RTECSNumber RG9700100
TSCA Yes