missing translation for 'onlineSavingsMsg'
Learn More

1-Iodoheptane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I0320500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0320500G 500 g
I032025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0320500G Supplier TCI America Supplier No. I0320500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4282-40-0
Molecular Formula C7H15I
Molecular Weight (g/mol) 226.101
MDL Number MFCD00001104
InChI Key LMHCYRULPLGEEZ-UHFFFAOYSA-N
Synonym heptyl iodide, n-heptyl iodide, heptane, 1-iodo, iodoheptane, n-heptyliodide, iodo heptane, 1-iodanylheptane, 1-iodomeptane, 1-iodo heptane, 1-iodo-heptane
PubChem CID 20274
IUPAC Name 1-iodoheptane
SMILES CCCCCCCI

Specifications

Melting Point -48°C
Boiling Point 204°C
Quantity 500 g
Formula Weight 226.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodoheptane (stabilized with Copper chip)
RTECSNumber MJ1050000
TSCA Yes