missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-3-methylbutane (stabilized with Na2S2O3) 99.0+%, TCI America™

Catalog No. I006125ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I006125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I006125ML Supplier TCI America Supplier No. I006125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 541-28-6
Molecular Formula C5H11I
Molecular Weight (g/mol) 198.047
MDL Number MFCD00039414
InChI Key BUZZUHJODKQYTF-UHFFFAOYSA-N
Synonym isoamyl iodide, isopentyl iodide, butane, 1-iodo-3-methyl, 3-methylbutyliodide, 1-iodo-3-methyl-butane, 1-jod-3-methylbutan, iso-amyliodide, unii-e1e5du187l, 1-jod-3-methylbutan czech, 1-iodo-3-methyl butane
PubChem CID 10924
IUPAC Name 1-iodo-3-methylbutane
SMILES CC(C)CCI

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 mL
UN Number 1993
Formula Weight 198.05
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Iodo-3-methylbutane (stabilized with Na2S2O3)
RTECSNumber EK4412000
TSCA Yes