missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylboronic acid pinacol ester, 98%

Catalog No. AAH2783114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2783114 25 g
AAH2783103 1 g
AAH2783106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2783114 Supplier Thermo Scientific Chemicals Supplier No. H2783114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86308-26-1 | C12H25BO2 | 212.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86308-26-1
Molecular Formula C12H25BO2
Molecular Weight (g/mol) 212.14
MDL Number MFCD08457665
InChI Key LDPMCSWZEGKWLC-UHFFFAOYSA-N
Synonym hexylboronic acid pinacol ester, 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 1,3,2-dioxaborolane, 2-hexyl-4,4,5,5-tetramethyl
PubChem CID 10921795
IUPAC Name 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCCCC

Specifications

Density 1.167
Flash Point 84°C (183°F)
Refractive Index 1.426
Quantity 25 g
Formula Weight 212.14
Percent Purity 98%
Chemical Name or Material 1-Hexylboronic acid pinacol ester
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only