missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl isothiocyanate, 97%

Catalog No. AAA1222314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1222314 25 g
AAA1222306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1222314 Supplier Thermo Scientific Chemicals Supplier No. A1222314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4404-45-9 | C7H13NS | 143.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4404-45-9
Molecular Formula C7H13NS
Molecular Weight (g/mol) 143.248
MDL Number MFCD00014445
InChI Key WXYAXKKXIGHXDS-UHFFFAOYSA-N
Synonym hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
PubChem CID 78120
IUPAC Name 1-isothiocyanatohexane
SMILES CCCCCCN=C=S

Specifications

Density 0.933
Boiling Point 216°C
Flash Point 90°C (194°F)
Odor Pungent
Refractive Index 1.4915
Quantity 25 g
UN Number UN2922
Beilstein 1751837
Sensitivity Moisture sensitive
Formula Weight 143.25
Percent Purity 97%
Chemical Name or Material 1-Hexyl isothiocyanate
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 224-549-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only