missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Iodide 98.0+%, TCI America™

Numéro de catalogue. p-7131135
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H16335G 5 g
H163325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H16335G Fournisseur TCI America Code fournisseur H16335G
Il en reste null

Identifiants chimiques

CAS 178631-05-5
Molecular Formula C10H19IN2
Molecular Weight (g/mol) 294.18
MDL Number MFCD12964976
InChI Key CZIUVCSYOGFUPH-UHFFFAOYSA-M
Synonym 1-hexyl-3-methylimidazolium iodide, 1-hexyl-3-methyl-1h-imidazol-3-ium iodide, 1-n-hexyl-3-methylimidazolium iodide, dsstox_cid_27927, dsstox_rid_82680, dsstox_gsid_47951, 1-methyl-3-hexylimidazolium iodide, 1-hexyl-3-me-thylimidazolium iodide, 1h-imidazolium,1-hexyl-3-methyl-,iodide, 1-hexyl-3-methylimidazolium iodide hplc
PubChem CID 12036651
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;iodide
SMILES CCCCCCN1C=C[N+](=C1)C.[I-]

Spécifications

Color Brown-Yellow
Quantity 5 g
Formula Weight 294.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Iodide