missing translation for 'onlineSavingsMsg'
Learn More

1-Hexyl-3-methylimidazolium Chloride 98.0+%, TCI America™

Catalog No. H10975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10975G 5 g
H109725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10975G Supplier TCI America Supplier No. H10975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 171058-17-6
Molecular Formula C10H19N2
Molecular Weight (g/mol) 167.28
MDL Number MFCD03093289
InChI Key RVEJOWGVUQQIIZ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
PubChem CID 2734163
ChEBI CHEBI:61340
IUPAC Name 3-hexyl-1-methyl-1H-imidazol-3-ium
SMILES CCCCCC[N+]1=CN(C)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 202.73
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Hexyl-3-methylimidazolium Chloride