missing translation for 'onlineSavingsMsg'
Learn More

1-Hexadecyne 90.0+%, TCI America™

Catalog No. H04335ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04335ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04335ML Supplier TCI America Supplier No. H04335ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-74-3
Molecular Formula C16H30
Molecular Weight (g/mol) 222.42
MDL Number MFCD00015084
InChI Key UCIDYSLOTJMRAM-UHFFFAOYSA-N
Synonym 1-hexadecyne, tetradecylacetylene, acmc-209nbc
PubChem CID 12396
IUPAC Name hexadec-1-yne
SMILES CCCCCCCCCCCCCCC#C

Specifications

Color Yellow
Boiling Point 284°C
Quantity 5 mL
Formula Weight 222.42
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Hexadecyne