missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Hexadécanéthiol 97,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 189°C |
| Quantity | 25 mL |
| Formula Weight | 258.51 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 1-Hexadecanethiol |
Sécurité et manipulation
| EINECSNumber | (2)-0464 |
| TSCA | Yes |