missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoroadamantane 98.0+%, TCI America™

Catalog No. F0511250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0511250MG 250 mg
F05111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0511250MG Supplier TCI America Supplier No. F0511250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 768-92-3
Molecular Formula C10H15F
Molecular Weight (g/mol) 154.228
InChI Key CPWSNJSGSXXVLD-UHFFFAOYSA-N
Synonym 1-Adamantyl Fluoride
PubChem CID 136608
IUPAC Name 1-fluoroadamantane
SMILES C1C2CC3CC1CC(C2)(C3)F

Specifications

Melting Point 257°C
Color White
Quantity 250 mg
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluoroadamantane