missing translation for 'onlineSavingsMsg'
Learn More

1-Ethynyl-4-(phenylethynyl)benzene 98.0+%, TCI America™

Numéro de catalogue. p-7129251
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E0967200MG 200 mg
E09671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E0967200MG Fournisseur TCI America Code fournisseur E0967200MG
Il en reste null

Identifiants chimiques

CAS 92866-00-7
Molecular Formula C16H10
Molecular Weight (g/mol) 202.26
MDL Number MFCD27665710
InChI Key WAUKYPQSFIENDE-UHFFFAOYSA-N
Synonym 4-Ethynyldiphenylacetylene, 1-(4-Ethynylphenyl)-2-phenylacetylene
PubChem CID 11275700
IUPAC Name 1-ethynyl-4-(2-phenylethynyl)benzene
SMILES C#CC1=CC=C(C=C1)C#CC1=CC=CC=C1

Spécifications

Melting Point 90°C
Color White
Quantity 200 mg
Formula Weight 202.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethynyl-4-(phenylethynyl)benzene