missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpyridinium Bromide 98.0+%, TCI America™

Catalog No. E017125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E017125G Supplier TCI America Supplier No. E017125G
Only null left

Chemical Identifiers

CAS 1906-79-2
Molecular Formula C7H10BrN
Molecular Weight (g/mol) 188.068
MDL Number MFCD00041999
InChI Key ABFDKXBSQCTIKH-UHFFFAOYSA-M
Synonym 1-ethylpyridinium bromide, 1-ethylpyridin-1-ium bromide, ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide, n-ethylpyridinium bromide, pyridinium, 1-ethyl-, bromide 1:1, pyridineethobromide, ethylpyridiniumbromide, 1-ethyl-pyridiniubromide, n-ethyl pyridinium bromide
PubChem CID 15927
IUPAC Name 1-ethylpyridin-1-ium;bromide
SMILES CC[N+]1=CC=CC=C1.[Br-]

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 188.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethylpyridinium Bromide