missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethylpyrazole 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 137°C |
| Quantity | 5 mL |
| UN Number | 1993 |
| Formula Weight | 96.13 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Ethylpyrazole |
Sécurité et manipulation
| TSCA | No |