missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-ethynylbenzene 97.0+%, TCI America™

Catalog No. E07495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07495G 5 g
E074925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07495G Supplier TCI America Supplier No. E07495G

Chemical Identifiers

CAS 40307-11-7
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00173887
InChI Key ZNTJVJSUNSUMPP-UHFFFAOYSA-N
Synonym 4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
PubChem CID 142425
IUPAC Name 1-ethyl-4-ethynylbenzene
SMILES CCC1=CC=C(C=C1)C#C

Specifications

Color Yellow
Boiling Point 192°C
Quantity 5 g
Formula Weight 130.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-4-ethynylbenzene