missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-eth-1-ynylbenzene, Thermo Scientific™

Chemical Identifiers

CAS 40307-11-7
Molecular Formula C10H10
Molecular Weight (g/mol) 130.19
MDL Number MFCD00173887
InChI Key ZNTJVJSUNSUMPP-UHFFFAOYSA-N
Synonym 4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
PubChem CID 142425
IUPAC Name 1-ethyl-4-ethynylbenzene
SMILES CCC1=CC=C(C=C1)C#C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BTB09906DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09906EA
View Documents Promotion Details
N/A 10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09906EB
View Documents Promotion Details
N/A 25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

40307-11-7
130.19
ZNTJVJSUNSUMPP-UHFFFAOYSA-N
142425
CCC1=CC=C(C=C1)C#C
C10H10
MFCD00173887
4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
1-ethyl-4-ethynylbenzene

Specifications

40307-11-7
100.0
97%
C10H10
MFCD00173887
ZNTJVJSUNSUMPP-UHFFFAOYSA-N
1-ethyl-4-ethynylbenzene
142425
97%
96.5
0.93g/mL
Amber glass bottle
HC2CC6H4CH2CH3
4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
CCC1=CC=C(C=C1)C#C
130.19
130.19
1-Ethyl-4-eth-1-ynylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents