missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Methyl Sulfate 98.0+%, TCI America™

Catalog No. E13085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13085G 5 g
E130825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E13085G Supplier TCI America Supplier No. E13085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 516474-01-4
Molecular Formula C7H14N2O4S
Molecular Weight (g/mol) 222.259
MDL Number MFCD08457697
InChI Key BXSDLSWVIAITRQ-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc
PubChem CID 16211409
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;methyl sulfate
SMILES CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-]

Specifications

Quantity 5 g
Formula Weight 222.26
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-3-methylimidazolium Methyl Sulfate