missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium Iodide 98.0+%, TCI America™

Catalog No. E05565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05565G 5 g
E055625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E05565G Supplier TCI America Supplier No. E05565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35935-34-3
Molecular Formula C6H11IN2
Molecular Weight (g/mol) 238.07
MDL Number MFCD03701101
InChI Key IKQCDTXBZKMPBB-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium iodide, 1-ethyl-3-methylimidazoliumiodide, 1-ethyl-3-methyl-1h-imidazol-3-ium iodide, acmc-209ik2, dsstox_cid_29137, dsstox_rid_83356, dsstox_gsid_49281, ksc495i0n, 1-ethyl-3-methyl imidazolium iodide salt
PubChem CID 11075478
IUPAC Name 1-ethyl-3-methyl-1H-imidazol-3-ium iodide
SMILES [I-].CCN1C=C[N+](C)=C1

Specifications

Melting Point 79°C
Color Yellow
Quantity 5 g
Formula Weight 238.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium Iodide