missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium hydrogen sulfate, 98%

Catalog No. AAH2723230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2723230 250 g
AAH2723218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2723230 Supplier Thermo Scientific Chemicals Supplier No. H2723230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 412009-61-1 | C6H12N2O4S | 208.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 412009-61-1
Molecular Formula C6H12N2O4S
Molecular Weight (g/mol) 208.23
MDL Number MFCD06798195
InChI Key HZKDSQCZNUUQIF-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium hydrogen sulfate, 1-ethyl-3-methylimidazolium hydrogensulfate, basionics™ ac 25, acmc-209jj9, dsstox_cid_29156, dsstox_rid_83375, dsstox_gsid_49300, 1-ethyl-3-methylimidazoliumhydrogensulfate, 1-methyl-3-ethylimidazolium hydrogensulfate, 1h-imidazolium,1-ethyl-3-methyl-,sulfate 1:1
PubChem CID 16217215
SMILES OS([O-])(=O)=O.CCN1C=C[N+](C)=C1

Specifications

Density 1.367
Quantity 250 g
UN Number UN3265
IUPAC Name 1-ethyl-3-methyl-1H-imidazol-3-ium hydrogen sulfate
Formula Weight 208.24
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium hydrogen sulfate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only