missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium hexafluorophosphate, 98+%

Numéro de catalogue. AAL1976204
Modifier l'affichage
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL1976204 2 g
AAL1976209 10 g
AAL1976218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAL1976204 Fournisseur Thermo Scientific Chemicals Code fournisseur L1976204
Il en reste null

CAS: 155371-19-0 | C6H11F6N2P | 256.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 155371-19-0
Molecular Formula C6H11F6N2P
Molecular Weight (g/mol) 256.132
MDL Number MFCD00216703
InChI Key DPDAKOVGQUGTHH-UHFFFAOYSA-N
Synonym 1-ethyl-3-methylimidazolium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, emimpf6, dsstox_cid_27897, dsstox_rid_82650, dsstox_gsid_47921, ksc174g8p, 1-ethyl-3-methylimidazol-3-ium hexafluorophosphate, 1-ethyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate, 1-ethyl-3-methylimidazolium hexafluorophosphate hplc
PubChem CID 2758873
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;hexafluorophosphate
SMILES CCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F

Spécifications

Melting Point 58°C to 62°C
Quantity 2 g
Sensitivity Hygroscopic
Formula Weight 256.13
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium hexafluorophosphate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only