missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium diethyl phosphate, 98%

Catalog No. AAH2740014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2740014 25 g
AAH2740006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2740014 Supplier Thermo Scientific Chemicals Supplier No. H2740014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 848641-69-0 | C10H21N2O4P | 264.26 g/mol

It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in improving enzymatic hydrolysis of wheat straw and in the enzymatic in situ saccharification of cellulose in aqueous-ionic liquid media.

Solubility
Fully miscible in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 848641-69-0
Molecular Formula C10H21N2O4P
Molecular Weight (g/mol) 264.26
MDL Number MFCD09953486
InChI Key HQWOEDCLDNFWEV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium diethyl phosphate, 1-ethyl-3-methylimidazolium diethylphosphate, dsstox_cid_27865, dsstox_rid_82618, dsstox_gsid_47889, emim dep, basionics™ lq 11, 1-ethyl-3-methylimidazolium-diethyl phosphate, 1-ethyl-3-methylimidazolium diethyl phosphate hplc
PubChem CID 53316406
IUPAC Name diethyl phosphate;1-ethyl-3-methylimidazol-3-ium
SMILES CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC

Specifications

Melting Point 19°C to 21°C
Density 1.157
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 264.26
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium diethyl phosphate
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only