missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium bromide, 98+%

Catalog No. AAL1976109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1976109 10 g
AAL1976118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1976109 Supplier Thermo Scientific Chemicals Supplier No. L1976109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 65039-08-9 | C6H11BrN2 | 191.07 g/mol

An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An ionic liquid, 1-ethyl-3-methylimidazolium bromide is used as reaction medium for the synthesis and crystallization of a coordination polymer, (EMI)[Cd(BTC)]. 1-ethyl-3-methylimidazolium bromide was used as a solvent in [emim] 2 [Cd 2 (btec)Br 2 ]. Form the diene complexation; carbonyl insertion; cycloaddition; reduction and deoxygenation.

Solubility
Soluble in water, dichloromethane, ethanol, acetonitrile. Insoluble in ethyl acetate, ether, and alkane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 65039-08-9
Molecular Formula C6H11BrN2
Molecular Weight (g/mol) 191.07
MDL Number MFCD03427610
InChI Key GWQYPLXGJIXMMV-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
PubChem CID 2734235
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;bromide
SMILES [Br-].CCN1C=C[N+](C)=C1

Specifications

Melting Point 70°C to 73°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,dichloromethane,ethanol,acetonitrile. Insoluble in ethyl acetate,ether,and alkane.
Formula Weight 191.07
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium bromide
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only