missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol 95.0+%, TCI America™

Catalog No. E0446500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0446500ML 500 mL
E044625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0446500ML Supplier TCI America Supplier No. E0446500ML

Chemical Identifiers

CAS 1569-02-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00067050
InChI Key JOLQKTGDSGKSKJ-UHFFFAOYNA-N
Synonym 1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
PubChem CID 15287
IUPAC Name 1-ethoxypropan-2-ol
SMILES CCOCC(C)O

Specifications

Color Colorless
Boiling Point 132°C
Quantity 500 mL
UN Number 1987
Formula Weight 104.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethoxy-2-propanol
EINECSNumber (2)-2424
RTECSNumber UB5250000
TSCA No