missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyne, 97%

Catalog No. AAL0297714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0297714 25 g
AAL0297706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0297714 Supplier Thermo Scientific Chemicals Supplier No. L0297714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 765-03-7 | C12H22 | 166.31 g/mol

1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.

Chemical Identifiers

CAS 765-03-7
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00008960
InChI Key ZVDBUOGYYYNMQI-UHFFFAOYSA-N
Synonym 1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
PubChem CID 69821
IUPAC Name dodec-1-yne
SMILES CCCCCCCCCCC#C

Specifications

Melting Point -19°C
Density 0.779
Boiling Point 214°C to 216°C
Flash Point 79°C (174°F)
Refractive Index 1.434
Quantity 25 g
Beilstein 1741320
Solubility Information Not miscible in water.
Formula Weight 166.31
Percent Purity 97%
Chemical Name or Material 1-Dodecyne
Show More Show Less
Hazard Category H227-H315-H318-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-134-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only