missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecanol 99.0+%, TCI America™
$32.28 - $53.16
Chemical Identifiers
| CAS | 112-53-8 |
|---|---|
| Molecular Formula | C12H26O |
| Molecular Weight (g/mol) | 186.34 |
| MDL Number | MFCD00004753 |
| InChI Key | LQZZUXJYWNFBMV-UHFFFAOYSA-N |
| Synonym | 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol |
| PubChem CID | 8193 |
| ChEBI | CHEBI:28878 |
| IUPAC Name | dodecan-1-ol |
| SMILES | CCCCCCCCCCCCO |
Chemical Identifiers
| C12H26O | |
| MFCD00004753 | |
| 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
| CHEBI:28878 | |
| CCCCCCCCCCCCO |
Specifications
| 112-53-8 | |
| 262°C | |
| MFCD00004753 | |
| 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol | |
| CCCCCCCCCCCCO | |
| 186.34 | |
| CHEBI:28878 | |
| ≥99.0% (GC) | |
| 1-Dodecanol |
| White | |
| C12H26O | |
| 3077 | |
| LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
| dodecan-1-ol | |
| 8193 | |
| 186.34 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0217&(2)-3704
RTECSNumber : JR5775000
TSCA : Yes