missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanol 99.0+%, TCI America™

Catalog No. D097825G
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D097825G 25 g
D0978400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D097825G Supplier TCI America Supplier No. D097825G

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Color White
Boiling Point 262°C
Quantity 25 g
UN Number 3077
Formula Weight 186.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Dodecanol
EINECSNumber (2)-0217&(2)-3704
RTECSNumber JR5775000
TSCA Yes