missing translation for 'onlineSavingsMsg'
Learn More

1-Decyne, 98%

Catalog No. AAA1117122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1117122 100 g
AAA1117106 5 g
AAA1117114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1117122 Supplier Thermo Scientific Chemicals Supplier No. A1117122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 764-93-2 | C10H18 | 138.254 g/mol

1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decyne has been used to stabilize ruthenium nanoparticles by forming ruthenium-vinylidene interfacial bonds. It is also used as primary and secondary intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with strong oxidizing agents. Store at 0-6°C.

Chemical Identifiers

CAS 764-93-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00009576
InChI Key ILLHQJIJCRNRCJ-UHFFFAOYSA-N
Synonym 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
PubChem CID 12997
ChEBI CHEBI:87322
IUPAC Name dec-1-yne
SMILES CCCCCCCCC#C

Specifications

Melting Point -44°C
Density 0.767
Boiling Point 174°C to 176°C
Flash Point 50°C (122°F)
Odor Petrol-like
Refractive Index 1.427
Quantity 100 g
UN Number UN3295
Beilstein 1236372
Solubility Information Not miscible in water.
Formula Weight 138.26
Percent Purity 98%
Chemical Name or Material 1-Decyne
Show More Show Less
Hazard Category H226-H315-H318
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 212-132-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only