missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)naphthalene, 95%

Catalog No. AC109455000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109455000 500 g Glass bottle
AC109450050 5 g Glass bottle
AC109451000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC109455000 Supplier Thermo Scientific Chemicals Supplier No. 109455000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 86-52-2 | C11H9Cl | 176.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-52-2
Molecular Formula C11H9Cl
Molecular Weight (g/mol) 176.65
MDL Number MFCD00004042
InChI Key XMWGTKZEDLCVIG-UHFFFAOYSA-N
Synonym 1-chloromethyl naphthalene, naphthalene, 1-chloromethyl, 1-naphthylmethyl chloride, 1-menaphthyl chloride, 1-chlormethyl naftalen, chloromethyl naphthalene, 1-chloro methylnaphthalene, alpha-naphthylmethyl chloride
PubChem CID 6845
IUPAC Name 1-(chloromethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CCl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 31°C to 31°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 291°C to 292°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range Remainder 2-chloromethylnaphtalene (HPLC)
Packaging Glass bottle
Refractive Index 1.6350 to 1.639
Linear Formula C10H7CH2Cl
Quantity 500 g
Beilstein 05, 566
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 176.65
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 1-(Chloromethyl)naphthalene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-678-2
RTECSNumber QJ2800000
TSCA TSCA

RUO – Research Use Only