missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroisoquinoline 98.0+%, TCI America™

Catalog No. C20975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20975G 5 g
C209725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20975G Supplier TCI America Supplier No. C20975G

Chemical Identifiers

CAS 19493-44-8
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.604
MDL Number MFCD00024134
InChI Key MSQCQINLJMEVNJ-UHFFFAOYSA-N
Synonym 1-chloro-isoquinoline, isoquinoline, 1-chloro, chloroisoquinoline, isoquinoline, chloro, 1-chloroisoquinone, 1-chloro isoquinoline, zlchem 1129, pubchem13906, acmc-1bowu, 1-chloroisoquinoline
PubChem CID 140539
IUPAC Name 1-chloroisoquinoline
SMILES C1=CC=C2C(=C1)C=CN=C2Cl

Specifications

Melting Point 36°C
Boiling Point 108°C
Quantity 5 g
Formula Weight 163.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloroisoquinoline
TSCA No
Recommended Storage Refrigerator